Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129177.4 | NP_001122649.1 | 4 - 5 | 598 | 77 | |
NM_064144.6 | NP_496545.1 | 4 - 5 | 613 | 77 | ||
GenBank mRNA | L39212.1 | - | 4 - 5 | 622 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |