Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rol-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064188.5 | NP_496589.1 | 2 - 3 | 186 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |