Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064255.3 | NP_496656.2 | 4 - 5 | 1338 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |