Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064320.6 | NP_496721.2 | 6 - 7 | 2360 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |