Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dct-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064354.7 | NP_496755.1 | 4 - 5 | 767 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |