Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zyx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267435.3 | NP_001254364.1 | 4 - 5 | 756 | 85 | |
NM_064375.8 | NP_496776.1 | 5 - 6 | 1008 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |