Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026956.5 | NP_001022127.1 | 1 - 2 | 189 | 92 | |
NM_001026957.6 | NP_001022128.1 | 1 - 2 | 182 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |