Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hpr-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064392.6 | NP_496793.1 | 2 - 3 | 185 | 74 | |
GenBank mRNA | AF076840.1 | - | 2 - 3 | 172 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |