Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hpr-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064392.6 | NP_496793.1 | 4 - 5 | 955 | 91 | |
GenBank mRNA | AF076840.1 | - | 4 - 5 | 942 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |