Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129186.3 | NP_001122658.1 | 3 - 4 | 551 | 82 | |
NM_001129188.4 | NP_001122660.1 | 3 - 4 | 551 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |