Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064455.6 | NP_496856.1 | 3 - 4 | 453 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |