Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hda-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064509.6 | NP_496910.1 | 2 - 3 | 257 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |