Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: imb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322761.3 | NP_001309574.1 | 3 - 4 | 1798 | 81 | |
NM_064586.4 | NP_496987.1 | 3 - 4 | 1798 | 81 | ||
NM_064587.5 | NP_496988.1 | 2 - 3 | 406 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |