Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: imb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322761.3 | NP_001309574.1 | 6 - 7 | 2598 | 69 | |
NM_064586.4 | NP_496987.1 | 6 - 7 | 2580 | 69 | ||
NM_064587.5 | NP_496988.1 | 5 - 6 | 1188 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |