Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nurf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026946.4 | NP_001022117.2 | 3 - 4 | 1034 | 113 | |
NM_001267499.2 | NP_001254428.1 | 3 - 4 | 1038 | 113 | ||
NM_001267500.3 | NP_001254429.1 | 3 - 4 | 1039 | 113 | ||
NM_001313609.3 | NP_001300538.1 | 2 - 3 | 669 | 113 | ||
NM_001313610.3 | NP_001300539.1 | 2 - 3 | 669 | 113 | ||
NM_001313611.3 | NP_001300540.1 | 3 - 4 | 1032 | 113 | ||
NM_001377899.3 | NP_001364539.1 | 3 - 4 | 1038 | 113 | ||
NM_001381656.2 | NP_001366940.1 | 3 - 4 | 1039 | 113 | ||
NM_001383991.1 | 4 - 5 | 949 | 113 | |||
NM_064593.4 | NP_496994.3 | 3 - 4 | 1039 | 113 | ||
NM_064594.9 | NP_496995.3 | 3 - 4 | 1038 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |