Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377900.1 | NP_001364609.1 | 13 - 14 | 4985 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |