Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267509.3 | NP_001254438.1 | 2 - 3 | 341 | 138 | |
NM_001377905.1 | NP_001364664.1 | 35 - 36 | 10098 | 138 | ||
NM_064643.10 | NP_497044.3 | 26 - 27 | 7838 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |