Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027317.8 | NP_001022488.1 | 13 - 14 | 4596 | 58 | |
NM_001267510.3 | NP_001254439.1 | 14 - 15 | 4007 | 58 | ||
NM_001267511.3 | NP_001254440.1 | 12 - 13 | 4226 | 58 | ||
NM_001267516.3 | NP_001254445.1 | 15 - 16 | 4784 | 58 | ||
NM_001267517.3 | NP_001254446.1 | 15 - 16 | 4784 | 58 | ||
NM_001350388.3 | NP_001337294.1 | 25 | 7151 | 58 | ||
NM_001377905.1 | NP_001364664.1 | 24 - 25 | 7044 | 58 | ||
NM_064643.10 | NP_497044.3 | 15 - 16 | 4784 | 58 | ||
NM_064644.6 | NP_497045.1 | 15 - 16 | 4874 | 58 | ||
NM_064645.6 | NP_497046.1 | 15 - 16 | 4875 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |