Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osta-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306651.3 | NP_001293580.1 | 2 - 3 | 411 | 65 | |
NM_064670.5 | NP_497071.1 | 5 - 6 | 797 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |