Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027210.7 | NP_001022381.1 | 6 - 7 | 1284 | 91 | |
NM_001027211.6 | NP_001022382.1 | 5 - 6 | 1155 | 91 | ||
NM_001306648.3 | NP_001293577.1 | 3 - 4 | 844 | 91 | ||
NM_001306649.3 | NP_001293578.1 | 3 - 4 | 853 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |