Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ver-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064761.2 | NP_497162.2 | 2 - 3 | 264 | 99 | |
GenBank mRNA | AF188751.1 | - | 2 - 3 | 264 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |