Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027405.6 | NP_001022576.1 | 2 - 3 | 132 | 60 | |
NM_001381707.2 | NP_001367353.1 | 1 - 2 | 75 | 60 | ||
NM_001381708.2 | NP_001367354.1 | 1 - 2 | 80 | 60 | ||
GenBank mRNA | U67966.1 | - | 1 - 2 | 78 | 60 | |
U67967.1 | - | 1 - 2 | 78 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |