Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064864.5 | NP_497265.1 | 2 - 3 | 479 | 101 | |
GenBank mRNA | U72883.1 | - | 2 - 3 | 435 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |