Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dac-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025958.4 | NP_001021129.1 | 3 - 4 | 556 | 74 | |
NM_001025959.5 | NP_001021130.1 | 3 - 4 | 473 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |