Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064966.2 | NP_497367.1 | 1 - 2 | 113 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |