Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065041.4 | NP_497442.2 | 2 - 3 | 185 | 86 | |
GenBank mRNA | EF471979.1 | - | 2 - 3 | 170 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |