Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027676.4 | NP_001022847.1 | 1 - 2 | 239 | 74 | |
NM_001373850.1 | NP_001359807.1 | 1 - 2 | 237 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |