Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129227.4 | NP_001122699.2 | 20 - 21 | 5244 | 150 | |
NM_001373851.2 | NP_001360184.1 | 8 | 2379 | 150 | ||
NM_065077.6 | NP_497478.2 | 19 - 20 | 5148 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |