Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306719.3 | NP_001293648.1 | 7 - 8 | 898 | 76 | |
NM_001381755.2 | NP_001367974.1 | 8 - 9 | 1040 | 76 | ||
NM_001381756.1 | NP_001367973.1 | 8 - 9 | 1315 | 76 | ||
NM_001381757.1 | NP_001367972.1 | 8 - 9 | 1325 | 76 | ||
NM_001384036.1 | 8 - 9 | 1038 | 76 | |||
GenBank mRNA | EU346943.1 | - | 7 - 8 | 898 | 76 | |
EU346944.1 | - | 7 - 8 | 898 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |