Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iffb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065135.8 | NP_497536.2 | 6 - 7 | 2197 | 70 | |
GenBank mRNA | DQ867020.1 | - | 6 - 7 | 2219 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |