Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phf-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027738.2 | NP_001022909.1 | 1 - 2 | 88 | 86 | |
GenBank mRNA | AF537097.1 | - | 1 - 2 | 81 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |