Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027461.4 | NP_001022632.1 | 12 - 13 | 2065 | 95 | |
NM_065139.8 | NP_497540.3 | 12 - 13 | 2065 | 95 | ||
GenBank mRNA | AY099352.1 | - | 12 - 13 | 2067 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |