Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smg-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381779.2 | NP_001367726.1 | 12 - 13 | 2488 | 109 | |
NM_065165.8 | NP_497566.3 | 13 - 14 | 2555 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |