Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381780.2 | NP_001368042.1 | 17 - 18 | 4016 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |