Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: feh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027735.5 | NP_001022906.1 | 6 - 7 | 802 | 89 | |
NM_001373858.2 | NP_001360128.1 | 4 - 5 | 643 | 89 | ||
NM_001384041.1 | 5 - 6 | 763 | 89 | |||
NM_065177.6 | NP_497578.4 | 6 - 7 | 807 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |