Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: feh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027735.5 | NP_001022906.1 | 8 - 9 | 1258 | 70 | |
NM_001373858.2 | NP_001360128.1 | 6 - 7 | 1099 | 70 | ||
NM_001384041.1 | 7 - 8 | 1219 | 70 | |||
NM_065177.6 | NP_497578.4 | 8 - 9 | 1263 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |