Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027766.4 | NP_001022937.1 | 8 - 9 | 2807 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |