Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065203.4 | NP_497604.2 | 4 - 5 | 861 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |