Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065262.6 | NP_497663.1 | 2 - 3 | 332 | 59 | |
GenBank mRNA | AB017190.1 | - | 2 - 3 | 274 | 59 | |
U65429.1 | - | 2 - 3 | 313 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |