Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065318.4 | NP_497719.1 | 7 - 8 | 792 | 73 | |
GenBank mRNA | AF082012.1 | - | 7 - 8 | 786 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |