Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136364.4 | NP_001129836.1 | 11 - 12 | 1711 | 76 | |
NM_065330.7 | NP_497731.1 | 11 - 12 | 1640 | 76 | ||
GenBank mRNA | AF083224.1 | - | 11 - 12 | 1662 | 76 | |
AY204167.1 | - | 10 - 11 | 1641 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |