Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065330.7 | NP_497731.1 | 3 - 4 | 211 | 98 | |
GenBank mRNA | AF083224.1 | - | 3 - 4 | 233 | 98 | |
AY204167.1 | - | 2 - 3 | 212 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |