Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyn-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065344.5 | NP_497745.1 | 3 - 4 | 422 | 92 | |
GenBank mRNA | U36581.1 | - | 3 - 4 | 415 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |