Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065358.5 | NP_497759.1 | 3 - 4 | 261 | 86 | |
GenBank mRNA | AF134806.1 | - | 3 - 4 | 263 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |