Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xbx-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065367.4 | NP_497768.2 | 3 - 4 | 758 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |