Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065401.5 | NP_497802.2 | 2 - 3 | 378 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |