Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065408.8 | NP_497809.1 | 1 - 2 | 98 | 108 | |
GenBank mRNA | X75564.1 | - | 109 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |