Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065408.8 | NP_497809.1 | 2 - 3 | 210 | 83 | |
GenBank mRNA | X75564.1 | - | 221 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |