Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065448.6 | NP_497849.1 | 4 - 5 | 544 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |