Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065479.8 | NP_497880.2 | 3 - 4 | 236 | 83 | |
GenBank mRNA | AY158691.1 | - | 1 - 2 | 144 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |